Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MFSD4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144856
Description
Immunogen sequence: SKERLLTCCP QRRPLLLSAD ELALETQPPE KEDASSLPPK FQSHLGHEDL FSCCQRKNLR GA Antigen sequence identity: Mouse 82%; Rat 81%.
MFSD4 gene ontology (GO) annotation includes glucose transmembrane transporter activity.
Specifications
MFSD4 | |
Polyclonal | |
Unconjugated | |
MFSD4A | |
major facilitator superfamily domain containing 4; major facilitator superfamily domain containing 4A; major facilitator superfamily domain-containing protein 4; Major facilitator superfamily domain-containing protein 4A; MFSD4; MFSD4A; UNQ3064; UNQ3064/PRO9894 | |
Rabbit | |
Affinity chromatography | |
RUO | |
148808 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N468 | |
MFSD4A | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 242-303. Recombinant protein control fragment (Product #RP-110015). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction