Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MFSD6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554581
Description
Immunogen sequence: PVPSSPVPIA TIDLVQQQTE DVMPRIEPRL PPKKTKHQEE QEDVNKPAWG VSSSPWVTFV YALYQIKEMM QLTRDNRASE IQPLQGTNEN RENSPAGRAQ PVPCETHSDP SRNQPSPDAA ASQTQTSPAH PSVDPC Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 83%.

Specifications
MFSD6 | |
Polyclonal | |
Unconjugated | |
MFSD6 | |
hMMR2; Macrophage MHC class I receptor 2 homolog; macrophage MHC receptor 2; major facilitator superfamily domain containing 6; major facilitator superfamily domain-containing protein 6; MFSD6; MMR2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54842 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZSS7 | |
MFSD6 | |
Recombinant protein corresponding to Human MFSD6. Recombinant protein control fragment (Product #RP-88605). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction