Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ MG 132
GREENER_CHOICESDP

Catalog No. 17485 Shop All Novus Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-485 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-485 Supplier R&D Systems™ Supplier No. 1748/5

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Proteasome and calpain inhibitor. Inhibits NF-κB activation

MG 132 is a potent cell-permeable inhibitor of proteasome (IC50 = 100 nM) and calpain (IC50 = 1.2 μM). MG 132 inhibits TNF-α-induced NF-κB activation and IκBα degradation. Induces neurite outgrowth in PC12 cells and has anticancer properties in vitro. MG 132 inhibits SARS-CoV-2 Mpro in vitro (IC50 = 3.9 μM) and inhibits viral replication. Also enhances adeno-associated viral transduction efficiency of human cell lines by around 50-fold.

Specifications

CAS 133407-82-6
Quantity 5 mg
Product Line Tocris
Product Type MG 132
Purity >95%
Molecular Weight (g/mol) 475.63
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.