Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MG-H1 Monoclonal Antibody (1H7G5)
GREENER_CHOICE

Catalog No. FERMA549069
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA549069 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA549069 Supplier Invitrogen™ Supplier No. MA549069
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MG-H1 Monoclonal Antibody for ICC/IF, Inhibition, Western Blot

MG-H1 is a gene that encodes a protein called Methylglyoxal-derived hydroimidazolone-1 (MG-H1). This protein is formed as a result of the reaction between methylglyoxal, a highly reactive molecule that is produced during various metabolic processes, and proteins in the body. MG-H1 is known to be involved in various pathological conditions, such as diabetes, atherosclerosis, and Alzheimer's disease. It is also believed to play a role in the aging process by contributing to the accumulation of damage in cells and tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MG-H1
Applications Inhibition Assays, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1H7G5
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene Accession No. P29466, P48729, Q8N752
Gene Alias methylglyoxal 5-hydro-5-methylimidazolones
Host Species Mouse
Immunogen N-alpha-acetyl-Ndelta-(5-hydro-5-methyl)-4-imidazolone.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122011, 1452, 834
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.