Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MGA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559934
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559934 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559934 Supplier Invitrogen™ Supplier No. PA559934
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLGDVKVEQQ KGFDNPEENS SEFPVTFKEE SKFELSGSKV MEQQSNLQPE AKEKECGDSL EKDRERWRKH LKGPLTRKCV GASQECKKEA DEQLIKETKT CQENSDVFQQ EQGISDLLGK SGITEDARVL KTECDSWSRI S Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 62%.

Functions as a dual-specificity transcription factor, regulating the expression of both MAX-network and T-box family target genes. Functions as a repressor or an activator. Binds to 5'-AATTTCACACCTAGGTGTGAAATT-3' core sequence and seems to regulate MYC-MAX target genes. Suppresses transcriptional activation by MYC and inhibits MYC-dependent cell transformation. Function activated by heterodimerization with MAX. This heterodimerization serves the dual function of both generating an E-box-binding heterodimer and simultaneously blocking interaction of a corepressor. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MGA
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MGA
Gene Accession No. Q8IWI9
Gene Alias AV312082; C130042M01Rik; C80739; D030062C11Rik; FLJ12634; KIAA0518; Kiaa4252; LOW QUALITY PROTEIN: MAX gene-associated protein; Mad5; maltase-glucoamylase (alpha-glucosidase); Max dimerization protein 5; MAX dimerization protein MGA; MAX gene associated; MAX gene-associated protein; MAX protein-associated protein-like protein; MAX-associated protein; MAX-interacting protein; MGA; MGA, MAX dimerization protein; MGA-like protein; MXD5; RGD1561597
Gene Symbols MGA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MGA. Recombinant protein control fragment (Product #RP-102486).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23269
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.