Learn More
Invitrogen™ MGARP Polyclonal Antibody

Description
Immunogen sequence: YKTVTSDQAKH TEHKTNLKEK TKAEIHPFQG EKENVAETEK ASSEAPEELI VEAEVVDAEE SPSATVVVIK EASACPGHVE AAPETTAVSA ETGPEVTDAA A Highest antigen sequence identity to the following orthologs - mouse 44%, rat 45%.
Specifications
Specifications
| Antigen | MGARP |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MGARP |
| Gene Accession No. | Q8TDB4 |
| Gene Alias | C4orf49; CESP1; CESP-1; Corneal endothelium-specific protein 1; GS3582; HUMMR; hypoxia up-regulated mitochondrial movement regulator protein; MGARP; mitochondria localized glutamic acid rich protein; mitochondria-localized glutamic acid-rich protein; OSAP; ovary-specific acidic protein; Protein MGARP |
| Gene Symbols | MGARP |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.