Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MGARP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA543494
Description
Peptide sequence: EAVTIDNDKD TTKNETSDEY AELEEENSPA ESESSAGDDL QEEASVGSEA Sequence homology: Human: 100%
MGARP Polyclonal antibody specifically detects MGARP in Human samples. It is validated for Western Blot.
Specifications
MGARP | |
Polyclonal | |
Unconjugated | |
MGARP | |
C4orf49; CESP1; CESP-1; Corneal endothelium-specific protein 1; GS3582; HUMMR; hypoxia up-regulated mitochondrial movement regulator protein; MGARP; mitochondria localized glutamic acid rich protein; mitochondria-localized glutamic acid-rich protein; OSAP; ovary-specific acidic protein; Protein MGARP | |
Rabbit | |
Affinity Chromatography | |
RUO | |
84709 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8TDB4 | |
MGARP | |
Synthetic peptide directed towards the C-terminal of human MGARP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction