Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ MGAT2, Mouse, Unlabeled, Clone: 4A5.2,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABS1703
Description
Alpha-1,6-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase (EC 2.4.1.143; UniProt Q10469; also known as Beta-1,2-N-acetylglucosaminyltransferase II, GlcNAc-T II, GNT-II, Mannoside acetylglucosaminyltransferase 2, MGAT2, N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyl-transferase II) is encoded by the MGAT2 (also known as CDG2A) gene (Gene ID 4247) in human. MGAT2 is a Golgi enzyme that catalyzes an essential step in the conversion of oligo-mannose to complex N-glycans. It possesses the typical glycosyltransferase domains: a short N-terminal cytoplasmic domain, a hydrophobic non-cleavable signal-anchor domain, and a C-terminal catalytic domain. It is highly expressed in small intestine in mice and humans. Mutations in MGTA2 gene are known to cause congenital disorders of glycosylation 2A, which is a multi-system disorder caused by defective glycoprotein biosynthesis. It leads to under-glycosylated levels of serum glycoproteins that result in defects in neuronal development, coagulation disorders, hypotonia, and immunodeficiency.
Specifications
MGAT2 | |
Monoclonal | |
Please refer to lot specific datasheet. | |
Purified mouse IgG2aκ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide. | |
Q10469 | |
GST-tagged internal fragment from the C-terminal half of human MGAT2. | |
100 μg | |
Signaling | |
NP_002399 | |
Human | |
Purified |
Western Blot | |
4A5.2 | |
Unconjugated | |
MGAT2, CDG2A | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Clone 4A5.2 targets an internal epitope within the C-terminal half of human MGAT2. | |
Stable for 1 year at 2°-8°C from date of receipt. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction