Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MGAT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566042
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566042 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566042 Supplier Invitrogen™ Supplier No. PA566042
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QVFFPFSIQLY PNEFPGSDPR DCPRDLPKNA ALKLGCINAE YPDSFGHYRE AKFSQTKHHW WWKLHFVWER Highest antigen sequence identity to the following orthologs - mouse 99%, rat 97%.

The product of this gene is a Golgi enzyme catalyzing an essential step in the conversion of oligomannose to complex N-glycans. The enzyme has the typical glycosyltransferase domains: a short N-terminal cytoplasmic domain, a hydrophobic non-cleavable signal-anchor domain, and a C-terminal catalytic domain. Mutations in this gene may lead to carbohydrate-deficient glycoprotein syndrome, type II. The coding region of this gene is intronless. Transcript variants with a spliced 5' UTR may exist, but their biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MGAT2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MGAT2
Gene Accession No. Q10469
Gene Alias AA407964; alpha-1,6-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase; Alpha-1,6-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase-like protein; beta-1,2-N-acetylglucosaminyltransferase II; beta-12-N-acetylglucosaminyltransferase II; CDG2A; CDGS2; CH73-301C19.1; GlcNAc-T II; GLCNACTII; GNT2; GNT-II; Mannoside acetylglucosaminyltransferase 2; mannosyl (alpha-1,6-)-glycoprotein beta-1,2-N-acetylglucosaminyltransferase; mgat2; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase II; UDP-N-acetylglucosamine:alpha-6-D-mannoside beta-1,2-N-acetylglucosaminyltransferase II; zgc:162268
Gene Symbols MGAT2
Host Species Rabbit
Immunogen Recombinant Human MGAT2. Recombinant protein control fragment (Product #RP-106732).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4247
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.