Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MGAT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597186
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597186 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597186 Supplier Invitrogen™ Supplier No. PA597186
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PFSIQLYPNE FPGSDPRDCP RDLPKNAALK LGCINAEYPD SFGHYREAKF SQTKHHWWWK LHFVWERVKI LRDYAGLILF LEEDHYLAPD FYHVFKKMWK LKQQECPECD VLSLGTYSAS RSFYGMADKV DVKTWKSTEH NMGLALTRNA YQKLIECTDT FCTYDDYNWD WTLQYLTVSC LPKFWKVLVP QIPRIFHAGD CGMHHKKTCR PSTQSAQIES LLNNNKQYMF PETLTISEKF TVVAISPPRK NGGWGDIRDH ELCKSYRRLQ; Positive Samples: K-562, Mouse kidney, Rat kidney; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein.

The product of this gene is a Golgi enzyme catalyzing an essential step in the conversion of oligomannose to complex N-glycans. The enzyme has the typical glycosyltransferase domains: a short N-terminal cytoplasmic domain, a hydrophobic non-cleavable signal-anchor domain, and a C-terminal catalytic domain. Mutations in this gene may lead to carbohydrate-deficient glycoprotein syndrome, type II. The coding region of this gene is intronless. Transcript variants with a spliced 5' UTR may exist, but their biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MGAT2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MGAT2
Gene Accession No. Q09326, Q10469, Q921V5
Gene Alias AA407964; alpha-1,6-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase; Alpha-1,6-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase-like protein; beta-1,2-N-acetylglucosaminyltransferase II; beta-12-N-acetylglucosaminyltransferase II; CDG2A; CDGS2; CH73-301C19.1; GlcNAc-T II; GLCNACTII; GNT2; GNT-II; Mannoside acetylglucosaminyltransferase 2; mannosyl (alpha-1,6-)-glycoprotein beta-1,2-N-acetylglucosaminyltransferase; mgat2; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase II; UDP-N-acetylglucosamine:alpha-6-D-mannoside beta-1,2-N-acetylglucosaminyltransferase II; zgc:162268
Gene Symbols MGAT2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 178-447 of human MGAT2 (NP_0023991).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217664, 4247, 94273
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.