Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MGAT4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552456
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552456 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552456 Supplier Invitrogen™ Supplier No. PA552456
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLKIHVNPPA EVSTSLKVYQ GHTLEKTYMG EDFFWAITPI AGDYILFKFD KPVNVESYLF HSGNQEHPGD ILLNTTVEVL PFKSEGLEIS KETKDKRLED GYFRIGKFEN GVAEGMVDPS LNPISAFRLS VIQ Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 95%.

This gene encodes a key glycosyltransferase that regulates the formation of tri- and multiantennary branching structures in the Golgi apparatus. The encoded protein, in addition to the related isoenzyme B, catalyzes the transfer of N-acetylglucosamine (GlcNAc) from UDP-GlcNAc in a beta-1,4 linkage to the Man-alpha-1,3-Man-beta-1,4-GlcNAc arm of R-Man-alpha-1,6(GlcNAc-beta-1,2-Man-alpha-1,3)Man-beta-1,4-GlcNAc-beta-1,4-GlcNAc-beta-1-Asn. The encoded protein may play a role in regulating the availability of serum glycoproteins, oncogenesis, and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MGAT4A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MGAT4A
Gene Accession No. Q9UM21
Gene Alias 2.4.1.145; 9530018I07Rik; alpha-1,3-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase A; Alpha-1,3-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase A soluble form; alpha-1,3-mannosyl-glycoprotein beta-1,4-N-acetylglucosaminyltransferase; glcNAc-T IVa; glcNAc-T-IVa; GnT-4a; GNT-IV; GNT-IVA; mannoside acetylglucosaminyltransferase 4, isoenzyme A; mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isoenzyme A; mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme A; Mgat4a; N-acetylglucosaminyltransferase IVa; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase IVa; UDP-GlcNAc:a-1,3-D-mannoside b-1,4-acetylglucosaminyltransferase IV; UDP-N-acetylglucosamine: alpha-1,3-D-mannoside beta-1,4-N-acetylglucosaminyltransferase IVa; UDP-N-acetylglucosamine:alpha1,3-d-mannoside beta1,4-N-acetylglucosaminyltransferase
Gene Symbols MGAT4A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MGAT4A. Recombinant protein control fragment (Product #RP-89705).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11320
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.