Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MGAT4A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572306
Description
MGAT4A Polyclonal Antibody for Western Blot
This gene encodes a key glycosyltransferase that regulates the formation of tri- and multiantennary branching structures in the Golgi apparatus. The encoded protein, in addition to the related isoenzyme B, catalyzes the transfer of N-acetylglucosamine (GlcNAc) from UDP-GlcNAc in a beta-1,4 linkage to the Man-alpha-1,3-Man-beta-1,4-GlcNAc arm of R-Man-alpha-1,6(GlcNAc-beta-1,2-Man-alpha-1,3)Man-beta-1,4-GlcNAc-beta-1,4-GlcNAc-beta-1-Asn. The encoded protein may play a role in regulating the availability of serum glycoproteins, oncogenesis, and differentiation.
Specifications
MGAT4A | |
Polyclonal | |
Unconjugated | |
MGAT4A | |
2.4.1.145; 9530018I07Rik; alpha-1,3-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase A; Alpha-1,3-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase A soluble form; alpha-1,3-mannosyl-glycoprotein beta-1,4-N-acetylglucosaminyltransferase; glcNAc-T IVa; glcNAc-T-IVa; GnT-4a; GNT-IV; GNT-IVA; mannoside acetylglucosaminyltransferase 4, isoenzyme A; mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isoenzyme A; mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme A; Mgat4a; N-acetylglucosaminyltransferase IVa; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase IVa; UDP-GlcNAc:a-1,3-D-mannoside b-1,4-acetylglucosaminyltransferase IV; UDP-N-acetylglucosamine: alpha-1,3-D-mannoside beta-1,4-N-acetylglucosaminyltransferase IVa; UDP-N-acetylglucosamine:alpha1,3-d-mannoside beta1,4-N-acetylglucosaminyltransferase | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
11320, 269181 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.45 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q812G0, Q9UM21 | |
MGAT4A | |
KLH conjugated synthetic peptide between 70-102 amino acids from human MGAT4A. | |
200 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction