Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MGAT4C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100611
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100611 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100611 Supplier Invitrogen™ Supplier No. PA5100611
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MGAT4C.

Glycosyltransferase that participates in the transfer of N-acetylglucosamine (GlcNAc) to the core mannose residues of N- linked glycans. Catalyzes the formation of the GlcNAcbeta1-4 branch on the GlcNAcbeta1-2Manalpha1-3 arm of the core structure of N-linked glycans. Essential for the production of tri- and tetra-antennary N-linked sugar chains. Does not catalyze the transfer of GlcNAc to the Manalpha1-6 arm to form GlcNAcBeta1-4Manalpha1-6 linkage ('GnT-VI' activity).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MGAT4C
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MGAT4C
Gene Accession No. Q9D306, Q9UBM8
Gene Alias 9130411I17Rik; Alpha-1,3-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase C; glcNAc-T IVc; GnT-IVc; GntIVh; hGnT-IV-H; mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme C; mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme C (putative); MGAT4 family member C; MGAT4 family, member C; MGAT4C; N-acetylglucosaminyltransferase IV homolog; N-acetylglucosaminyltransferase IVc; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase IVc; RGD1307871; UDP-N-acetylglucosamine: alpha-1,3-D-mannoside beta-1,4-N-acetylglucosaminyltransferase IVc; UDP-N-acetylglucosamine:a-1,3-D-mannoside beta-1,4-N-acetylglucosaminyltransferase IV; UDP-N-acetylglucosamine:a-1,3-D-mannoside beta-1,4-N-acetylglucosaminyltransferase IV-homolog
Gene Symbols MGAT4C
Host Species Rabbit
Immunogen A synthesized peptide derived from human MGAT4C(Accession Q9UBM8), corresponding to amino acid residues L295-L345.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25834, 299756, 67569
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.