Learn More
Invitrogen™ MGAT4C Polyclonal Antibody

Description
Immunogen sequence: KNVIRFKPSL FQHMGYYSSY KGTENKLKDD DFEEESFDIP DNPPASLYTN MNVFENYEAS KAYSSVDEYF WGKPPSTGDV FVIVFENPII IKKIKVNTGT EDRQNDILHH GALDVGENVM PSKQRRQCST YLRLGEFKNG NFEMSGVNQK IPFDIHCMRI YVTKTQKEWL IIRSISIWTS; Positive Samples: U-87MG; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein.
Specifications
Specifications
| Antigen | MGAT4C |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 3.26 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MGAT4C |
| Gene Accession No. | Q9UBM8 |
| Gene Alias | 9130411I17Rik; Alpha-1,3-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase C; glcNAc-T IVc; GnT-IVc; GntIVh; hGnT-IV-H; mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme C; mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme C (putative); MGAT4 family member C; MGAT4 family, member C; MGAT4C; N-acetylglucosaminyltransferase IV homolog; N-acetylglucosaminyltransferase IVc; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase IVc; RGD1307871; UDP-N-acetylglucosamine: alpha-1,3-D-mannoside beta-1,4-N-acetylglucosaminyltransferase IVc; UDP-N-acetylglucosamine:a-1,3-D-mannoside beta-1,4-N-acetylglucosaminyltransferase IV; UDP-N-acetylglucosamine:a-1,3-D-mannoside beta-1,4-N-acetylglucosaminyltransferase IV-homolog |
| Gene Symbols | MGAT4C |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.