Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MGAT4C Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548410
Description
Predicted to react with monkey, mouse and pig based on sequence homology.
Glycosyltransferase that participates in the transfer of N-acetylglucosamine (GlcNAc) to the core mannose residues of N- linked glycans. Catalyzes the formation of the GlcNAcbeta1-4 branch on the GlcNAcbeta1-2Manalpha1-3 arm of the core structure of N-linked glycans. Essential for the production of tri- and tetra-antennary N-linked sugar chains. Does not catalyze the transfer of GlcNAc to the Manalpha1-6 arm to form GlcNAcBeta1-4Manalpha1-6 linkage (′GnT-VI′ activity).
Specifications
MGAT4C | |
Polyclonal | |
Unconjugated | |
MGAT4C | |
Alpha-1, 3-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase C, N-acetylglucosaminyltransferase IV homolog, hGnT-IV-H, N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase IVc, GlcNAc-T IVc, GnT-IVc, N-acetylglucosaminyltransferase IVc, UDP-N-acetylglucosamine: alpha-1, 3-D-mannoside beta-1, 4-N-acetylglucosaminyltransferase IVc, MGAT4C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25834 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.44 mg/mL | |
PBS with 0.09% sodium azide | |
Q9UBM8 | |
MGAT4C | |
KLH conjugated synthetic peptide between 303-332 amino acids from the C-terminal region of human MGAT4C. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction