Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MGAT5 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA548637

Catalog No. PA548637


Only null left
Add to Cart

Description

Description

MGAT5 Polyclonal Antibody for Western Blot, Flow

Catalyzes the addition of N-acetylglucosamine in beta 1- 6 linkage to the alpha-linked mannose of biantennary N-linked oligosaccharides. It is one of the most important enzymes involved in the regulation of the biosynthesis of glycoprotein oligosaccharides.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MGAT5
Polyclonal
Unconjugated
MGAT5
4930471A21Rik; 5330407H02Rik; AI480971; alpha-1,6-mannosyl-glycoprotein 6-beta-N-acetylglucosaminyltransferase; alpha-1,6-mannosylglycoprotein 6-beta-N-acetylglucosaminyltransferase A; Alpha-mannoside beta-1,6-N-acetylglucosaminyltransferase; Alpha-mannoside beta-1,6-N-acetylglucosaminyltransferase V; beta1,6N-acetylglucosaminyltransferase V; EC 2.4.1.155; GGNT5; GlcNAc-T V; GlcNAc-TV; GNT-V; GNT-VA; GNT-VGNT-VA; mannoside acetylglucosaminyltransferase 5; mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase; Mgat5; N-acetylglucosaminyltransferase V; N-acetylglucosaminyl-transferase V; Secreted alpha-1,6-mannosylglycoprotein 6-beta-N-acetylglucosaminyltransferase A; Secreted beta-1,6-N-acetylglucosaminyltransferase V; Secreted GNT-V
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
4249
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q09328
MGAT5
KLH conjugated synthetic peptide between 652-680 amino acids from the C-terminal region of human MGAT5.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.