Learn More
Invitrogen™ MGAT5 Polyclonal Antibody

Description
Immunogen sequence: HGQVMWPPLS ALQVKLAEPG QSCKQVCQES QLICEPSFFQ HLNKDKDMLK YKVTCQSSEL AKDILVPSFD PKNKHCVFQG DLLLFSCAGA HPRHQRVCPC RDFIKGQVAL CKDCL; Positive Samples: LO2, 293T, Raji, Jurkat; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein.
Specifications
Specifications
| Antigen | MGAT5 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.19 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | MGAT5 |
| Gene Accession No. | Q09328, Q8R4G6 |
| Gene Alias | 4930471A21Rik; 5330407H02Rik; AI480971; alpha-1,6-mannosyl-glycoprotein 6-beta-N-acetylglucosaminyltransferase; alpha-1,6-mannosylglycoprotein 6-beta-N-acetylglucosaminyltransferase A; Alpha-mannoside beta-1,6-N-acetylglucosaminyltransferase; Alpha-mannoside beta-1,6-N-acetylglucosaminyltransferase V; beta1,6N-acetylglucosaminyltransferase V; EC 2.4.1.155; GGNT5; GlcNAc-T V; GlcNAc-TV; GNT-V; GNT-VA; GNT-VGNT-VA; mannoside acetylglucosaminyltransferase 5; mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase; Mgat5; N-acetylglucosaminyltransferase V; N-acetylglucosaminyl-transferase V; Secreted alpha-1,6-mannosylglycoprotein 6-beta-N-acetylglucosaminyltransferase A; Secreted beta-1,6-N-acetylglucosaminyltransferase V; Secreted GNT-V |
| Gene Symbols | MGAT5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.