Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MGAT5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986848
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6848 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6848 Supplier Bioss Supplier No. BS5841R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Catalyzes the addition of N-acetylglucosamine in beta 1- 6 linkage to the alpha-linked mannose of biantennary N-linked oligosaccharides. It is one of the most important enzymes involved in the regulation of the biosynthesis of glycoprotein oligosaccharides.

Specifications

Antigen MGAT5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MGAT5
Gene Accession No. Q09328
Gene Alias 4930471A21Rik; 5330407H02Rik; AI480971; Alpha-1,6-mannosylglycoprotein 6-beta-N-acetylglucosaminyltransferase A; Alpha-mannoside beta-1,6-N-acetylglucosaminyltransferase; Alpha-mannoside beta-1,6-N-acetylglucosaminyltransferase V; beta1,6N-acetylglucosaminyltransferase V; EC 2.4.1.155; GGNT5; GlcNAc-T V; GlcNAc-TV; GNT-V; GNT-VA; GNT-VGNT-VA; Mannoside acetylglucosaminyltransferase 5; mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase; Mgat5; N-acetylglucosaminyl-transferase V; Secreted alpha-1,6-mannosylglycoprotein 6-beta-N-acetylglucosaminyltransferase A; Secreted beta-1,6-N-acetylglucosaminyltransferase V; Secreted GNT-V
Gene Symbols MGAT5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MGAT5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4249
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.