Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MGAT5B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100612
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100612 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100612 Supplier Invitrogen™ Supplier No. PA5100612
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MGAT5B.

Glycosyltransferase that acts on alpha-linked mannose of N-glycans and O-mannosyl glycans. Catalyzes the transfer of N-acetylglucosamine (GlcNAc) to the beta 1-6 linkage of the mannose residue of GlcNAcbeta1,2-Manalpha on both the alpha1,3- and alpha1,6-linked mannose arms in the core structure of N-glycan. Also acts on the GlcNAcbeta1,2-Manalpha1-Ser/Thr moiety, forming a 2,6-branched structure in brain O-mannosyl glycan. Plays an active role in modulating integrin and laminin-dependent adhesion and migration of neuronal cells via its activity in the O-mannosyl glycan pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MGAT5B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MGAT5B
Gene Accession No. Q3V5L5, Q765H6
Gene Alias Alpha-1,6-mannosylglycoprotein 6-beta-N-acetylglucosaminyltransferase B; alpha-mannoside beta-1,6-N-acetylglucosaminyltransferase B; beta(1,6)-N-acetylglucosaminyltransferase V; beta1,6-N-acetylglucosaminyltransferase IX (GnT-IX); C330018B01; GlcNAc-T Vb; GnT-IX; GNT-Vb; hGnTVb; KIAA2008; mannoside acetylglucosaminyltransferase 5, isoenzyme B; mannoside acetylglucosaminyltransferase 5B; mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase, isoenzyme B; mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase, isozyme B; Mgat5b; mGnTVB; N-acetylglucosaminyltranferase VB; N-acetylglucosaminyltransferase IX; N-acetylglucosaminyl-transferase Vb; UDP-N-acetylglucosamine: alpha1,6-D-mannoside beta1,6-N-acetylglucosaminyltransferase b
Gene Symbols MGAT5B
Host Species Rabbit
Immunogen A synthesized peptide derived from human MGAT5B(Accession Q3V5L5), corresponding to amino acid residues D136-A186.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146664, 268510, 303693
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.