Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MGAT5B Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305134100UL
Description
MGAT5B Polyclonal specifically detects MGAT5B in Mouse samples. It is validated for Western Blot.Specifications
MGAT5B | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
146664 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
alpha-1,6-mannosylglycoprotein 6-beta-N-acetylglucosaminyltransferase B, Alpha-mannoside beta-1,6-N-acetylglucosaminyltransferase B, beta(1,6)-N-acetylglucosaminyltransferase V, EC 2.4.1.-, EC 2.4.1.155, FLJ25132, GlcNAc-T Vb, GnT-IX, GNT-Vb, hGnTVb, isoenzyme B, isozyme B, KIAA2008, Mannoside acetylglucosaminyltransferase 5B, mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase, N-acetylglucosaminyltransferase IX, N-acetylglucosaminyl-transferase Vb | |
Recombinant fusion protein containing a sequence corresponding to amino acids 616-790 of human MGAT5B (NP_653278.2). | |
100 μL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction