Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MGC DAU Control Sets

Catalog No. p-6162723
Click to view available options
Type:
Clinical DAU Control Set
Optional DAU Control Set
Primary DAU Control Set
Select DAU Control Set
Specialty DAU Control Set
This item is not returnable. View return policy

Catalog No. 23-278-254

Supplier: Thermo Scientific™ 100200

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Monitor Thermo Scientific™ CEDIA™ and Thermo Scientific™ DRI™ drugs of abuse testing assay performance with Thermo Scientific MGC DAU Control Sets.

Monitor Thermo Scientific™ CEDIA™ and Thermo Scientific™ DRI™ drugs of abuse testing assay performance with Thermo Scientific MGC DAU Control Sets. These controls contain high and low level controls at drug concentrations 25% below and above screening assay cutoffs to ensure quality laboratory results.

Key Features
  • Multi-constituent
  • For use in the validation of drug of abuse assays in human urine
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Detectable Analytes Amphetamine, Secobarbital, Oxazepam, Benzoylecgonine, Benzoylecgonine, d-Methamphetamine, EDDP, Methadone, Methaqualone, Morphine, Nitrazepam, Opiate 2000 (Morphine), Phencyclidine (PCP), Propoxyphene (PPX), Secobarbital
Sample Type Urine
Type Primary DAU Control Set
Includes Amphetamine: 750ng/mL and 1250ng/mL; Barbiturate: 150ng/mL and 250ng/mL; Benzodiazepine: 150ng/mL and 250ng/mL; Cocaine: 225ng/mL and 375ng/mL; Methadone: 225ng/mL and 375ng/mL; Methadone Metabolite (EDDP): 750ng/mL and 1250ng/mL; Methaqualone: 225ng/mL a
Quantity 3 x 5 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.