Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MGC12965 Monoclonal Antibody (OTI4C7)
GREENER_CHOICE

Catalog No. PIMA526965
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526965 Supplier Invitrogen™ Supplier No. MA526965
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MGC12965 Monoclonal Antibody for Western Blot, IHC (P)

MGC12965 functions in DNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MGC12965
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4C7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LOC100287896
Gene Accession No. 0
Gene Alias LOC100287896; MGC12965; uncharacterized LOC100287896; uncharacterized protein LOC100287896
Gene Symbols LOC100287896
Host Species Mouse
Immunogen Full length human recombinant protein of MGC12965 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100287896
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.