Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MGC20410 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | MGC20410 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17926820
![]() |
Novus Biologicals
NBP17926820UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179268
![]() |
Novus Biologicals
NBP179268 |
100 μL |
Each for $487.50
|
|
|||||
Description
MGC20410 Polyclonal specifically detects MGC20410 in Human samples. It is validated for Western Blot.Specifications
MGC20410 | |
Polyclonal | |
Rabbit | |
NP_612465 | |
116071 | |
Synthetic peptide directed towards the N terminal of human BATF2. Peptide sequence MHLCGGNGLLTQTDPKEQQRQLKKQKNRAAAQRSRQKHTDKADALHQQHE. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
basic leucine zipper transcription factor, ATF-like 2, basic leucine zipper transcriptional factor ATF-like 2, B-ATF-2, MGC20410, SARI, suppressor of AP-1 regulated by IFN | |
BATF2 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title