Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRR22 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548186
Description

Specifications
PRR22 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q8IZ63 | |
PRR22 | |
KLH conjugated synthetic peptide between 368-397 amino acids from the C-terminal region of human MGC24975. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
PRR22 | |
Proline-rich protein 22, PRR22 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
163154 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction