Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MGC4172 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23068625UL
Description
MGC4172 Polyclonal specifically detects MGC4172 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MGC4172 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Q6UWP2 | |
DHRS11 | |
This antibody was developed against a recombinant protein corresponding to amino acids: LVQQGLKVVGCARTVGNIEELAAECKSAGYPGTLIPYRCDLSNEEDILSMFSAIRSQHSGVDICINNAGL | |
25 μL | |
Stem Cell Markers | |
79154 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ARPG836, dehydrogenase/reductase (SDR family) member 11, dehydrogenase/reductase SDR family member 11, FLJ39232, MGC4172, SDR24C1, short chain dehydrogenase/reductase family 24C, member 1, short-chain dehydrogenase/reductase | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction