Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MGC87631 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544154
Description
Peptide sequence: VDKRDRKKSI QQLVPEYKEK QTPESLPQNN NPAAPSQAEG GEGGVACGTV Sequence homology: Human: 100%.
The function of this protein remains unknown.
Specifications
| MGC87631 | |
| Polyclonal | |
| Unconjugated | |
| CCDC144NL | |
| CCDC144NL; coiled-coil domain containing 144 family, N-terminal like; putative coiled-coil domain-containing protein 144 N-terminal-like | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 339184 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6NUI1 | |
| CCDC144NL | |
| Synthetic peptide directed towards the middle region of human MGC87631. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction