Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MgcRacGAP/RACGAP1 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP147267H
Description
MgcRacGAP/RACGAP1 Polyclonal antibody specifically detects MgcRacGAP/RACGAP1 in Human samples. It is validated for Western BlotSpecifications
MgcRacGAP/RACGAP1 | |
Polyclonal | |
Western Blot | |
GTPase activating protein, HsCYK-4, ID-GAP, KIAA1478, Male germ cell RacGap, MGCRACGAP, MgcRacGAPrac GTPase-activating protein 1, Rac GTPase activating protein 1 | |
The immunogen for this product maps to a region between residue 150 and 200 of human Rac GTPase-Activating Protein 1 using the numbering given in entry NP_001119575.1 (GeneID 29127). | |
0.1 mL | |
Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
29127 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction