Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mgea5 Recombinant Rabbit Monoclonal Antibody (8D12)
GREENER_CHOICE

Catalog No. FEMA549925
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA549925 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA549925 Supplier Invitrogen™ Supplier No. MA549925
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Mgea5 Recombinant Monoclonal Antibody for Western Blot, ELISA

This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This protein forms homo- or hetero-dimers with other Ikaros family members, and is thought to function predominantly in early hematopoietic development. Multiple transcript variants encoding different isoforms have been found for this gene, but the biological validity of some variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mgea5
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 8D12
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Oga
Gene Accession No. O60502
Gene Alias 2810009A20Rik; 4833427O07Rik; 5830447M11Rik; AA408215; beta-hexosaminidase; Beta-N-acetylglucosaminidase; beta-N-acetylhexosaminidase; Bifunctional protein NCOAT; Hexc; Hy5; hyaluronidase in meningioma; KIAA0679; MEA5; meningioma expressed antigen 5 (hyaluronidase); meningioma-expressed antigen 5; Mgea5; mKIAA0679; N-acetyl-beta-D-glucosaminidase; N-acetyl-beta-glucosaminidase; NCOAT; Nuclear cytoplasmic O-GlcNAcase and acetyltransferase; OGA; O-GLCNACASE; Protein O-GlcNAcase
Gene Symbols MGEA5, Oga
Host Species Rabbit
Immunogen A synthesized peptide derived from human MGEA5.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10724
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.