Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MGLL Monoclonal Antibody (OTI4D1), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503067S


Description
MGLL Monoclonal antibody specifically detects MGLL in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
MGLL | |
Monoclonal | |
0.62 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8IYD1 | |
Mouse | |
Affinity chromatography | |
RUO | |
23708 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2b |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI4D1 | |
Unconjugated | |
GSPT2 | |
AA589436; HUK5; HU-K5; lysophospholipase homolog; Lysophospholipase-like; MAGL; Mgl; Mgl2; MGLL; Monoacylglycerol lipase; monoglyceride lipase | |
Full length human recombinant protein of human GSPT2 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction