Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mGluR1/5 Glutamate Receptor Monoclonal Antibody (S75-33), QED Bioscience™
SDP

Catalog No. 502278221
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8221 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8221 Supplier Leinco Technologies Inc Supplier No. 56526100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

mGluR1/5 Glutamate Receptor Monoclonal antibody specifically detects mGluR1/5 Glutamate Receptor in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot

Specifications

Antigen mGluR1/5 Glutamate Receptor
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone S75-33
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene Grm5
Gene Accession No. P23385, P31424, P41594, P97772, Q13255, Q3UVX5
Gene Alias 4930455H15Rik; 6430542K11Rik; AI850523; ENSMUSG00000075319; G protein coupled receptor family C group 1 member A; G protein coupled receptor, family C, group 1, member A; G protein coupled receptor, family C, group 1, member E; G protein-coupled receptor GRM5; G protein-coupled receptor, family C, group 1, member A; G protein-coupled receptor, family C, group 1, member E; Glu5R; glutamate metabotropic receptor 1; glutamate metabotropic receptor 5; glutamate receptor; glutamate receptor, metabotropic 1; glutamate receptor, metabotropic 5; Gm10828; GPRC1A; Gprc1e; Grm1; GRM5; metabotropic glutamate receptor (mGluR5); metabotropic glutamate receptor 1; Metabotropic glutamate receptor 5; metabotropic glutamate receptor 5b; metabotropic glutamate receptor subtype 5b; metabotropic glutamate receptor type 5; MGLU1; mGlu5; mGluR1; mGluR5; mGluR5b; mGlurR1a; nmf373; PPP1R85; PPP1R86; protein phosphatase 1, regulatory subunit 85; protein phosphatase 1, regulatory subunit 86; rcw; SCAR13; wobl
Host Species Mouse
Immunogen Fusion protein corresponding to aa 824-1203 (cytoplasmic C-terminus) of rat mGluR5b (accession no. NP_058708.1).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108071, 14816, 24414, 24418, 2911, 2915
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.