Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mGluR1 Antibody (511601), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB48361Z
Description
mGluR1 Monoclonal antibody specifically detects mGluR1 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
mGluR1 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
511601 | |
Flow Cytometry, CyTOF-ready | |
glutamate receptor, metabotropic 1, GRM1A, metabotropic glutamate receptor 1, mGluR1, MGLUR1A, MGLUR1GPRC1AmGlu1 | |
Chinese hamster ovary cell line CHO-derived recombinant human mGluR1a, Met1-Ser522, Accession # Q13255 | |
0.1 mL | |
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision | |
2911 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction