Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mGluR1 Antibody (5E12D8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | mGluR1 |
---|---|
Clone | 5E12D8 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61827
![]() |
Novus Biologicals
NBP261827 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
mGluR1 Monoclonal specifically detects mGluR1 in Human samples. It is validated for Western Blot, ELISA.Specifications
mGluR1 | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
glutamate receptor, metabotropic 1, GRM1A, metabotropic glutamate receptor 1, mGluR1, MGLUR1A, MGLUR1GPRC1AmGlu1 | |
GRM1 | |
IgG1 | |
Protein G purified |
5E12D8 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
GPCR, Metabotropic Glutamate Receptors, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision | |
2911 | |
Purified recombinant fragment of human GRM1 (AA: extra 387-486) expressed in E. Coli. | |
Primary | |
132.4 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title