Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ mGluR1/mGluR5 Monoclonal Antibody (N75/3), APC
GREENER_CHOICE

Catalog No. PIMA545580
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545580 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545580 Supplier Invitrogen™ Supplier No. MA545580
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45580 was sufficient for detection of mGluR1/5 glutamate receptor in 20 μg of rat brain membrane lysate and assayed by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 130kDa (both mGluR1 and mGluR5). This antibody was formerly sold as clone S75-3.

may play a role in synaptic transmission; may interact with mGluR1, another group I metabotropic glutamate receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mGluR1/mGluR5
Applications Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N75/3
Concentration 1 mg/mL
Conjugate APC
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene Grm1
Gene Accession No. P23385, P31424, P41594, P97772, Q13255, Q3UVX5
Gene Alias 4930455H15Rik; 6430542K11Rik; AI850523; ENSMUSG00000075319; G protein coupled receptor family C group 1 member A; G protein coupled receptor, family C, group 1, member A; G protein coupled receptor, family C, group 1, member E; G protein-coupled receptor GRM5; G protein-coupled receptor, family C, group 1, member A; G protein-coupled receptor, family C, group 1, member E; Glu5R; glutamate metabotropic receptor 1; glutamate metabotropic receptor 5; glutamate receptor; glutamate receptor, metabotropic 1; glutamate receptor, metabotropic 5; Gm10828; GPRC1A; Gprc1e; Grm1; GRM5; metabotropic glutamate receptor (mGluR5); metabotropic glutamate receptor 1; Metabotropic glutamate receptor 5; metabotropic glutamate receptor 5b; metabotropic glutamate receptor subtype 5b; metabotropic glutamate receptor type 5; MGLU1; mGlu5; mGluR1; mGluR5; mGluR5b; mGlurR1a; nmf373; PPP1R85; PPP1R86; protein phosphatase 1, regulatory subunit 85; protein phosphatase 1, regulatory subunit 86; rcw; SCAR13; wobl
Gene Symbols Grm1, Grm5
Host Species Mouse
Immunogen Fusion protein amino acids 824-1203 (cytoplasmic C-terminus) of rat mGluR5b.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108071, 14816, 24414, 24418, 2911, 2915
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.