Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MGLUR1 + MGLUR5 Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss
SDP

Catalog No. 501981870
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1870 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1870 Supplier Bioss Supplier No. BS12014R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

may play a role in synaptic transmission; may interact with mGluR1, another group I metabotropic glutamate receptor.

Specifications

Antigen MGLUR1 + MGLUR5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Grm1
Gene Accession No. P23385, P31424, P41594, P97772, Q13255, Q3UVX5
Gene Alias G protein coupled receptor family C group 1 member A; glutamate receptor, metabotropic 1; glutamate receptor, metabotropic 5; Gprc1a; Gprc1e; Grm1; Grm5; metabotropic glutamate receptor (mGluR5); Metabotropic glutamate receptor 1; Metabotropic glutamate receptor 5; metabotropic glutamate receptor 5b; Mglur1; Mglur5; mGlurR1a
Gene Symbols Grm1, Grm5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MGLUR1 + MGLUR5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108071, 14816, 24414, 24418, 2911, 2915
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.