Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ mGluR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. OPA104060
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OPA104060 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OPA104060 Supplier Invitrogen™ Supplier No. OPA104060
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is specific to MGluR1a and does not cross-react with other splice variants.

Glutamate is the main excitatory neurotransmitter in the brain. For many years it had been considered to act only on the ligand-gated receptor channels-termed NMDA, AMPA and kainite receptors that are involved in the fast excitatory synaptic transmission. Recently, glutamate has been shown to regulate enzymes producing second messengers via specific receptors coupled to G-proteins. These receptors are called metabotropic glutamate receptors. In expression systems, Group-I receptors stimulate phospholipase C as revealed by an increase in phosphoinositide turnover and calcium release from internal stores. Group-II and -III receptors are coupled to the inhibition of adenylyl cyclase. The Group-I receptors include mGluR1a and mGluR5. The Group-II receptors in include mGluR2 and mGluR3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mGluR1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene Grm1
Gene Accession No. P23385, P97772
Gene Alias 4930455H15Rik; ENSMUSG00000075319; G protein coupled receptor family C group 1 member A; G protein coupled receptor, family C, group 1, member A; G protein-coupled receptor, family C, group 1, member A; glutamate metabotropic receptor 1; glutamate receptor; glutamate receptor, metabotropic 1; Gm10828; GPRC1A; GRM1; GRM1A; Metabotropic glutamate receptor 1; MGLU1; MGLUR1; mGluR1a; nmf373; PPP1R85; protein phosphatase 1, regulatory subunit 85; rcw; SCAR13; wobl
Gene Symbols Grm1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminal region of rat GRM1 conjugated to KLH.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14816, 24414
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.