Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mGluR2 Antibody (455310), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4676S
Description
mGluR2 Monoclonal antibody specifically detects mGluR2 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
mGluR2 | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
455310 | |
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
GLUR2, glutamate metabotropic receptor 2, glutamate receptor, metabotropic 2, GPRC1Bmetabotropic glutamate receptor 2, mGlu2, mGluR2, MGLUR2glutamate receptor homolog | |
Mouse myeloma cell line NS0-derived recombinant human mGluR2, Glu19-Ser498, Accession # Q14416 | |
0.1 mL | |
Apoptosis, Cancer, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Tumor Suppressors, Vision | |
2912 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction