Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mGluR2 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP168855
Description
mGluR2 Polyclonal specifically detects mGluR2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
mGluR2 | |
Polyclonal | |
Western Blot 1 - 3 ug/ml, Immunohistochemistry 3 - 5 ug/ml, Immunohistochemistry-Paraffin 3 - 5 ug/ml, Peptide ELISA Detection limit 1:16000 | |
NP_000830.2 | |
GRM2 | |
Peptide with sequence C-KETAPERREV corresponding to internal region according to NP_001123535.1. | |
0.1 mg | |
Apoptosis, Cancer, GPCR, Metabotropic Glutamate Receptors, Neuronal Cell Markers, Neuroscience, Neurotransmission, Tumor Suppressors, Vision | |
2912 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
GLUR2, glutamate metabotropic receptor 2, glutamate receptor, metabotropic 2, GPRC1Bmetabotropic glutamate receptor 2, mGlu2, mGluR2, MGLUR2glutamate receptor homolog | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is expected to recognize both isoforms (NP_000830.2; NP_001123535.1). | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction