Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ mGluR2/mGluR3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA130151

Catalog No. PIPA130151


Only null left
Add to Cart

Description

Description

PA1-30151 detects mGluR2 + mGluR3 from rat samples. Sequence is 100% conserved between mGluR2 and mGluR3. Predicted to react with human and mouse.

The mGluR proteins (metabotropic Glutamate Receptors) are members of the G-protein-coupled receptor family and are functionally and pharmacologically distinct from the GluR proteins (ionotropic Glutamate Receptors). The eight currently known mGluR proteins are mediated by two G-proteins with opposing regulation of adenylate cyclase pathways. The activities of mGluR1 and mGluR5 are mediated by a G-protein that activates a phosphatidylinositol-calcium second messenger system and generates a calcium-activated chloride current. The remainder of the eight sub-types of mGluR have an activity mediated by a G-protein that inhibits adenylate cyclase activity. mGluR-3, which may interact with GRASP, acts as a receptor for glutamate.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

mGluR2/mGluR3
Polyclonal
Unconjugated
Grm2
0710001G23Rik; 4930441L02Rik; G protein coupled receptor, family C, group 1, member B; G protein-coupled receptor, family C, group 1, member B; G protein-coupled receptor, family C, group 1, member C; GLUR2; GLUR3; glutamate metabotropic receptor 2; glutamate metabotropic receptor 3; glutamate receptor; glutamate receptor homolog; glutamate receptor, metabotropic 2; glutamate receptor, metabotropic 3; Gprc1b; GPRC1C; GRM2; Grm3; M mGlu3; Metabotropic glutamate receptor 2; metabotropic glutamate receptor 3; mGlu2; mGlu3; mGluR2; MGLUR3; mGluR7
Rabbit
Affinity Chromatography
RUO
24415, 24416
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry, Western Blot, Immunocytochemistry
1 mg/mL
PBS with 0.02% sodium azide
P31421, P31422
Grm2, Grm3
Rat mGlurR2/3 residues 860-872 (NGREVVDSTTSSL).
100 μg
Primary
Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.