Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ mGluR2/mGluR3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599485
Description
Antibody detects endogenous levels of total GluR2/3.
The mGluR proteins (metabotropic Glutamate Receptors) are members of the G-protein-coupled receptor family and are functionally and pharmacologically distinct from the GluR proteins (ionotropic Glutamate Receptors). The eight currently known mGluR proteins are mediated by two G-proteins with opposing regulation of adenylate cyclase pathways. The activities of mGluR1 and mGluR5 are mediated by a G-protein that activates a phosphatidylinositol-calcium second messenger system and generates a calcium-activated chloride current. The remainder of the eight sub-types of mGluR have an activity mediated by a G-protein that inhibits adenylate cyclase activity. mGluR-3, which may interact with GRASP, acts as a receptor for glutamate.
Specifications
mGluR2/mGluR3 | |
Polyclonal | |
Unconjugated | |
Grm2 | |
0710001G23Rik; 4930441L02Rik; G protein coupled receptor, family C, group 1, member B; G protein-coupled receptor, family C, group 1, member B; G protein-coupled receptor, family C, group 1, member C; GLUR2; GLUR3; glutamate metabotropic receptor 2; glutamate metabotropic receptor 3; glutamate receptor; glutamate receptor homolog; glutamate receptor, metabotropic 2; glutamate receptor, metabotropic 3; Gprc1b; GPRC1C; GRM2; Grm3; M mGlu3; Metabotropic glutamate receptor 2; metabotropic glutamate receptor 3; mGlu2; mGlu3; mGluR2; MGLUR3; mGluR7 | |
Rabbit | |
Affinity chromatography | |
RUO | |
108068, 108069, 24415, 24416, 2912, 2913 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P31421, P31422, Q14416, Q14832, Q14BI2, Q9QYS2 | |
Grm2, Grm3 | |
A synthesized peptide derived from human GRM3(Accession Q14832), corresponding to amino acid residues S847-L879. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction