Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human mGluR2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB4676SP
This item is not returnable.
View return policy
Description
mGluR2 Monoclonal specifically detects mGluR2 in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
mGluR2 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells | |
Q14416 | |
GRM2 | |
Mouse myeloma cell line NS0-derived recombinant human mGluR2 Glu19-Ser498 Accession # Q14416 | |
25 μg | |
Primary | |
Detects human mGluR2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with the N-terminal regions of recombinant human mGluR1, -3, -4, -5, -7, or -8 is observed. | |
Human | |
Purified |
Western Blot, Flow Cytometry | |
455310 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
GLUR2, glutamate metabotropic receptor 2, glutamate receptor, metabotropic 2, GPRC1B, GPRC1Bmetabotropic glutamate receptor 2, GRM2, mGlu2, mGluR2, MGLUR2glutamate receptor homolog, mGluR7 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2912 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction