Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mGluR2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA123573
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA123573 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA123573 Supplier Thermo Scientific Supplier No. PA123573

Rabbit Polyclonal Antibody

PA1-23573 detects Metabotropic Glutamate Receptor 2 in Human samples.

Glutamate is the main excitatory neurotransmitter in the brain. For many years it had been considered to act only on the ligand-gated receptor channels-termed NMDA, AMPA and kainite receptors that are involved in the fast excitatory synaptic transmission. Recently, glutamate has been shown to regulate enzymes producing second messengers via specific receptors coupled to G-proteins. These receptors are called metabotropic glutamate receptors. In expression systems, Group-I receptors stimulate phospholipase C as revealed by an increase in phosphoinositide turnover and calcium release from internal stores. Group-II and -III receptors are coupled to the inhibition of adenylyl cyclase. The Group-I receptors include mGluR1a and mGluR5. The Group-II receptors in include mGluR2 and mGluR3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mGluR2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GRM2
Gene Accession No. Q14416
Gene Alias 4930441L02Rik; G protein coupled receptor, family C, group 1, member B; G protein-coupled receptor, family C, group 1, member B; GLUR2; glutamate metabotropic receptor 2; glutamate receptor; glutamate receptor homolog; glutamate receptor, metabotropic 2; GPRC1B; Grm2; M mGlu2; Metabotropic; Metabotropic glutamate receptor 2; mGlu2; MGLUR2; mGluR7
Gene Symbols GRM2
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from N-terminal extracellular domain of human mGluR2
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2912
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.