Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mGluR3 Antibody (7A5A6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$560.50
Specifications
Antigen | mGluR3 |
---|---|
Clone | 7A5A6 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61843
![]() |
Novus Biologicals
NBP261843 |
0.1 mL |
Each of 1 for $560.50
|
|
|||||
Description
mGluR3 Monoclonal specifically detects mGluR3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
mGluR3 | |
1 mg/mL | |
Monoclonal | |
Purified | |
GPCR, Metabotropic Glutamate Receptors, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision | |
2913 | |
Purified recombinant fragment of human GRM3 (AA: extra 433-576) expressed in E. Coli. | |
Primary | |
99 kDa |
7A5A6 | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
Unconjugated | |
Mouse | |
GLUR3, glutamate receptor, metabotropic 3, GPRC1Cglutamate metabotropic receptor 3, GRM3, mGlu3, mGluR3, MGLUR3metabotropic glutamate receptor 3 | |
GRM3 | |
IgG2a | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title