Learn More
mGluR3 Polyclonal Antibody, Invitrogen™
Description
Recommended positive controls: Rat Cerebellar and cortical Membrane.
Specifications
Specifications
| Antigen | mGluR3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | tris glycine with 1% BSA and 0.1% sodium azide; pH 7.8 |
| Gene | GRM3 |
| Gene Accession No. | P31422, Q14832, Q9QYS2 |
| Gene Alias | 0710001G23Rik; G protein-coupled receptor, family C, group 1, member C; GLUR3; glutamate metabotropic receptor 3; glutamate receptor; glutamate receptor, metabotropic 3; Gprc1c; GRM3; M mGlu3; Metabotropic glutamate receptor 3; mGlu3; mGluR3 |
| Gene Symbols | GRM3 |
| Host Species | Rabbit |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.