Learn More
Invitrogen™ mGluR4 Polyclonal Antibody

Description
mGluR4 Polyclonal Antibody detects endogenous levels of mGluR4 protein.
Specifications
Specifications
| Antigen | mGluR4 |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | GRM4 |
| Gene Accession No. | P31423, Q14833, Q68EF4 |
| Gene Alias | G protein-coupled receptor GRM4; G protein-coupled receptor, family C, group 1, member D; glutamate metabotropic receptor 4; glutamate receptor; glutamate receptor, metabotropic 4; GPRC1D; GRM4; Metabotropic glutamate receptor 4; mGlu4; MGLUR4 |
| Gene Symbols | GRM4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.