Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mGluR5/1 Antibody (S75-3), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222424AF700
Description
mGluR5/1 Monoclonal specifically detects mGluR5/1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
metabotropic Glutamate Receptor 5/1 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GRM5 | |
Fusion protein amino acids 824-1203 (cytoplasmic C-terminus) of rat mGluR5b | |
0.1 mL | |
Primary | |
Detects approx 130kDa (both mGluR1 and mGluR5). | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
S75-3 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
glutamate receptor, metabotropic 5, GPRC1EMGLUR5metabotropic glutamate receptor 5, mGlu5, mGluR5 | |
Mouse | |
Protein G purified | |
Metabotropic Glutamate Receptors, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
2915 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction