Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ mGluR5 Recombinant Rabbit Monoclonal Antibody (ST51-00)
GREENER_CHOICE

Catalog No. PIMA532155
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532155 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532155 Supplier Invitrogen™ Supplier No. MA532155
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

GRK1 is a member of the guanine nucleotide protein-coupled (G protein) receptor kinase subfamily of Serine/Threonine kinase family. GRK1 phosphorylates rhodopsin and initiates its deactivation. G protein mediated phosphorylation promotes the binding of arrestin, which removes the receptor from the G-protein and terminates the receptor signal chain. GRK1 is farnesylated to promote membrane association. Studies have shown that GRK1 has been shown to be inhibited by recoverin, a Calcium binding protein. Diseases associated with GRK1 protein dysfunction include Oguchi Disease-2 and Hereditary Night Blindness.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mGluR5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot, Western Blot
Classification Recombinant Monoclonal
Clone ST51-00
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% Glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene Grm5
Gene Accession No. P31424, P41594, Q3UVX5
Gene Alias 6430542K11Rik; A830007B11Rik; AI850523; D16Ium24; D16Ium24e; G protein coupled receptor, family C, group 1, member E; G protein-coupled receptor GRM5; G protein-coupled receptor, family C, group 1, member E; Glu5R; Glur5; Glur-5; Glurbeta1; glutamate metabotropic receptor 5; glutamate receptor; glutamate receptor, metabotropic 5; GPRC1A; GPRC1E; GRM 1; GRM 5; GRM1; GRM1 alpha; GRM1A; Grm5; metabotropic 1; metabotropic glutamate receptor (mGluR5); metabotropic glutamate receptor 5; metabotropic glutamate receptor 5b; metabotropic glutamate receptor subtype 5b; metabotropic glutamate receptor type 5; MGC25101; mGlu1; mGlu5; mGluR; mGluR1; mGluR1 alpha; mGluR1A; MGLUR5; mGluR5a; mGluR5b; PPP1R86; protein phosphatase 1, regulatory subunit 86
Gene Symbols Grm5
Host Species Rabbit
Immunogen Synthetic peptide within Human GRM5 aa 1,163-1,212.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108071, 24418, 2915
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.