Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mGluR6 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300189G
Description
mGluR6 Polyclonal specifically detects mGluR6 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
mGluR6 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
CSNB1B, glutamate receptor, metabotropic 6, GPRC1Fmetabotropic glutamate receptor 6, mGlu6, mGluR6, MGLUR6DKFZp686H1993 | |
Rabbit | |
96 kDa | |
0.1 mL | |
Amino Acids Drugs and other small molecules, GPCR, Metabotropic Glutamate Receptors, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction, Vision | |
2916 | |
Mouse, Rat | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GRM6 | |
C-terminus of rat metabotropic Glutamate Receptor 6 (mGluR6) [Uniprot: P35349] | |
Affinity Purified | |
RUO | |
Primary | |
Mouse and rat. Reactivity has been poor on human brain tissue. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction