Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mGluR6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
$206.50 - $499.50
Specifications
Antigen | mGluR6 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Frozen |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB300189SS
![]() |
Novus Biologicals
NB300189SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB300189
![]() |
Novus Biologicals
NB300189 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
mGluR6 Polyclonal specifically detects mGluR6 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
mGluR6 | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Frozen | |
Polyclonal | |
Rabbit | |
Amino Acids Drugs and other small molecules, GPCR, Metabotropic Glutamate Receptors, Neuronal Cell Markers, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Vision | |
Tris-Glycine with 0.02% Sodium Azide | |
2916 | |
C-terminus of rat metabotropic Glutamate Receptor 6 (mGluR6) [Uniprot: P35349] | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. | |
96 kDa |
1 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
Unconjugated | |
RUO | |
Mouse, Rat | |
CSNB1B, glutamate receptor, metabotropic 6, GPRC1Fmetabotropic glutamate receptor 6, mGlu6, mGluR6, MGLUR6DKFZp686H1993 | |
GRM6 | |
IgG | |
Affinity Purified | |
Mouse and rat. Reactivity has been poor on human brain tissue. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title