Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ mGluR6 Monoclonal Antibody (4C11E12)
GREENER_CHOICE

Catalog No. PIMA531752
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531752 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531752 Supplier Invitrogen™ Supplier No. MA531752
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31752 has been tested in indirect ELISA.

Glutamate receptors constitute the principal excitatory neurotransmitter receptors in brain. Two classes of glutamate receptors exist: Ionotropic receptors, and metabotropic receptors. Ionotropic glutamate receptors are oligomeric complexes of various subunits (GluR17, NMDA13, KAI2), which comprise ligand-gated calcium channels. Metabotropic glutamate receptors are G-protein coupled receptors, which when activated can activate phopholipases or adenylylcyclase, depending on the neuron.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mGluR6
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4C11E12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene GRM6
Gene Accession No. O15303
Gene Alias BC021919; CSNB1B; G protein-coupled receptor GRM6; G protein-coupled receptor, family C, group 1, member F; glutamate metabotropic receptor 6; glutamate receptor; glutamate receptor, metabotropic 6; Gm3; GPRC1F; Grm6; Metabotropic glutamate receptor 6; mGlu6; MGLUR6; nerg1; nob2; nob3; nob4
Gene Symbols GRM6
Host Species Mouse
Immunogen Purified recombinant fragment of human GRM6 (AA: extra 480-585) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2916
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.