Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ mGluR6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA132783
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA132783 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA132783 Supplier Invitrogen™ Supplier No. PA132783
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-32783 detects GPRC1F from human samples.

Glutamate receptors constitute the principal excitatory neurotransmitter receptors in brain. Two classes of glutamate receptors exist: Ionotropic receptors, and metabotropic receptors. Ionotropic glutamate receptors are oligomeric complexes of various subunits (GluR17, NMDA13, KAI2), which comprise ligand-gated calcium channels. Metabotropic glutamate receptors are G-protein coupled receptors, which when activated can activate phopholipases or adenylylcyclase, depending on the neuron.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mGluR6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GRM6
Gene Accession No. O15303, P35349, Q5NCH9, Q863I4
Gene Alias BC021919; CSNB1B; G protein-coupled receptor GRM6; G protein-coupled receptor, family C, group 1, member F; glutamate metabotropic receptor 6; glutamate receptor; glutamate receptor, metabotropic 6; Gm3; GPRC1F; Grm6; Metabotropic glutamate receptor 6; mGlu6; MGLUR6; nerg1; nob2; nob3; nob4
Gene Symbols GRM6
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from N-terminal extracellular domain of human GRM6 / MGLUR6.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009501, 100768692, 102162790, 108072, 24419, 2916, 481452, 529656
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.